Assay Details
| Gene Symbol: LOC102176145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059878.1 | XP_017915367.1 | 6 - 7 | 857 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102176145| Gene Symbol: LOC102176145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059878.1 | XP_017915367.1 | 6 - 7 | 857 | 73 | |
Set Membership: |
|