Assay Details
| Gene Symbol: ZEB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062536.1 | XP_017918025.1 | 5 - 6 | 721 | 56 | |
| XM_018062541.1 | XP_017918030.1 | 5 - 6 | 792 | 56 | ||
| XM_018062545.1 | XP_017918034.1 | 5 - 6 | 1038 | 56 | ||
| XM_018062548.1 | XP_017918037.1 | 5 - 6 | 756 | 56 | ||
| XM_018062553.1 | XP_017918042.1 | 5 - 6 | 688 | 56 | ||
| XM_018062559.1 | XP_017918048.1 | 5 - 6 | 830 | 56 | ||
| GenBank mRNA | JO396936.2 | - | 3 - 4 | 495 | 56 | |
| JO582266.1 | - | 5 - 6 | 1181 | 56 | ||
| JT725614.1 | - | 4 - 5 | 615 | 56 | ||
More Information
Set Membership: |
|

ZEB2
Genomic Map