Assay Details
| Gene Symbol: GPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064565.1 | XP_017920054.1 | 9 - 10 | 1170 | 59 | |
| XM_018064566.1 | XP_017920055.1 | 10 - 11 | 1086 | 59 | ||
| GenBank mRNA | JO503490.2 | - | 10 - 11 | 1022 | 59 | |
| JT747023.1 | - | 4 - 5 | 1051 | 59 | ||
More Information
Set Membership: |
|

GPS2
Genomic Map