Assay Details
| Gene Symbol: LOC102185402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065861.1 | XP_017921350.1 | 13 - 14 | 1589 | 98 | |
| XM_018065862.1 | XP_017921351.1 | 13 - 14 | 1584 | 98 | ||
More Information
Set Membership: |
|

LOC102185402
Genomic Map