Assay Details
| Gene Symbol: LOC102185402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065861.1 | XP_017921350.1 | 2 - 3 | 439 | 69 | |
| XM_018065862.1 | XP_017921351.1 | 2 - 3 | 434 | 69 | ||
More Information
Set Membership: |
|

LOC102185402
Genomic Map