Assay Details
| Gene Symbol: LOC102185402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065861.1 | XP_017921350.1 | 9 - 10 | 1044 | 112 | |
| XM_018065862.1 | XP_017921351.1 | 9 - 10 | 1039 | 112 | ||
| GenBank mRNA | JO321395.2 | - | 2 - 3 | 226 | 112 | |
More Information
Set Membership: |
|

LOC102185402
Genomic Map