Assay Details
| Gene Symbol: LOC102185402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065861.1 | XP_017921350.1 | 11 - 12 | 1324 | 79 | |
| XM_018065862.1 | XP_017921351.1 | 11 - 12 | 1319 | 79 | ||
| GenBank mRNA | JO321395.2 | - | 4 - 5 | 506 | 79 | |
More Information
Set Membership: |
|

LOC102185402
Genomic Map