Assay Details
| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039647.1 | XP_017895136.1 | 2 - 3 | 545 | 62 | |
| XM_018039648.1 | XP_017895137.1 | 2 - 3 | 545 | 62 | ||
More Information
Set Membership: |
|

MALT1
Genomic Map