Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974707.2 | XP_013830161.1 | 5 - 6 | 243 | 63 | |
| GenBank mRNA | JO609035.2 | - | 4 - 5 | 359 | 63 | |
| JO609036.2 | - | 5 - 6 | 229 | 63 | ||
More Information
Set Membership: |
|

IL32
Genomic Map