Assay Details
Gene Symbol: ERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675706.2 | XP_005675763.2 | 3 - 4 | 499 | 58 | |
XM_018052850.1 | XP_017908339.1 | 5 - 6 | 714 | 58 | ||
XM_018052857.1 | XP_017908346.1 | 5 - 6 | 659 | 58 | ||
XM_018052858.1 | XP_017908347.1 | 5 - 6 | 714 | 58 | ||
XM_018052859.1 | XP_017908348.1 | 3 - 4 | 499 | 58 | ||
XM_018052860.1 | XP_017908349.1 | 5 - 6 | 714 | 58 | ||
XM_018052862.1 | XP_017908351.1 | 2 - 3 | 280 | 58 | ||
GenBank mRNA | JT738465.1 | - | 9 - 10 | 2163 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
