Assay Details
| Gene Symbol: DNAJB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049274.1 | XP_017904763.1 | 5 - 6 | 879 | 76 | |
| XM_018049275.1 | XP_017904764.1 | 4 - 5 | 525 | 76 | ||
More Information
Set Membership: |
|

DNAJB14
Genomic Map