Assay Details
| Gene Symbol: HSH2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682142.3 | XP_005682199.2 | 3 - 4 | 495 | 66 | |
| XM_005682143.3 | XP_005682200.2 | 3 - 4 | 371 | 66 | ||
| XM_018051332.1 | XP_017906821.1 | 2 - 3 | 245 | 66 | ||
More Information
Set Membership: |
|

HSH2D
Genomic Map