Assay Details
| Gene Symbol: ADGRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051678.1 | XP_017907167.1 | 11 - 12 | 1604 | 85 | |
| GenBank mRNA | KA311502.1 | - | 3 - 4 | 199 | 85 | |
| Gene Symbol: LOC102180663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051687.1 | XP_017907176.1 | 17 - 18 | 2240 | 85 | |
More Information
Set Membership: |
|

ADGRE2
Genomic Map