Assay Details
| Gene Symbol: LOC102171874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684175.3 | XP_005684232.3 | 1 - 2 | 660 | 84 | |
| XM_018052502.1 | XP_017907991.1 | 1 - 2 | 512 | 84 | ||
| GenBank mRNA | JO424743.1 | - | 1 - 2 | 564 | 84 | |
More Information
Set Membership: |
|

LOC102171874
Genomic Map