Assay Details
| Gene Symbol: CKAP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686664.2 | XP_005686721.2 | 1 - 2 | 96 | 62 | |
| XM_018055329.1 | XP_017910818.1 | 1 - 2 | 50 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018055329.1 transcripts.
Set Membership: |
|

CKAP2L
Genomic Map