Assay Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055944.1 | XP_017911433.1 | 6 - 7 | 641 | 60 | |
| XM_018055945.1 | XP_017911434.1 | 6 - 7 | 751 | 60 | ||
| GenBank mRNA | JO412476.2 | - | 6 - 7 | 648 | 60 | |
| JO584636.1 | - | 6 - 7 | 686 | 60 | ||
More Information
Set Membership: |
|

DOLPP1
Genomic Map