Assay Details
| Gene Symbol: RSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687977.3 | XP_005688034.1 | 8 - 9 | 903 | 61 | |
| GenBank mRNA | JO503512.2 | - | 1 - 2 | 570 | 61 | |
| KA311083.1 | - | 8 - 9 | 862 | 61 | ||
More Information
Set Membership: |
|

RSU1
Genomic Map