Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689064.3 | XP_005689121.2 | 5 - 6 | 1180 | 66 | |
| GenBank mRNA | JO430577.2 | - | 5 - 6 | 1162 | 66 | |
More Information
Set Membership: |
|

RDH10
Genomic Map