Assay Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058378.1 | XP_017913867.1 | 3 - 4 | 445 | 89 | |
| GenBank mRNA | JO453700.2 | - | 1 - 2 | 212 | 89 | |
| JO579323.1 | - | 3 - 4 | 539 | 89 | ||
More Information
Set Membership: |
|

FABP5
Genomic Map