Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061142.1 | XP_017916631.1 | 1 - 2 | 524 | 70 | |
| XM_018061143.1 | XP_017916632.1 | 1 - 2 | 524 | 70 | ||
| XM_018061144.1 | XP_017916633.1 | 1 - 2 | 523 | 70 | ||
| XM_018061145.1 | XP_017916634.1 | 1 - 2 | 529 | 70 | ||
| GenBank mRNA | JT710174.1 | - | 1 - 2 | 339 | 70 | |
More Information
Set Membership: |
|

BICDL1
Genomic Map