Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061142.1 | XP_017916631.1 | 5 - 6 | 1188 | 97 | |
| XM_018061143.1 | XP_017916632.1 | 6 - 7 | 1200 | 97 | ||
| XM_018061144.1 | XP_017916633.1 | 5 - 6 | 1187 | 97 | ||
| XM_018061145.1 | XP_017916634.1 | 6 - 7 | 1205 | 97 | ||
| GenBank mRNA | JT710174.1 | - | 5 - 6 | 1005 | 97 | |
More Information
Set Membership: |
|

BICDL1
Genomic Map