Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061142.1 | XP_017916631.1 | 8 - 9 | 1685 | 58 | |
| XM_018061143.1 | XP_017916632.1 | 8 - 9 | 1553 | 58 | ||
| XM_018061144.1 | XP_017916633.1 | 7 - 8 | 1540 | 58 | ||
| XM_018061145.1 | XP_017916634.1 | 9 - 10 | 1702 | 58 | ||
| GenBank mRNA | JT710174.1 | - | 7 - 8 | 1354 | 58 | |
More Information
Set Membership: |
|

BICDL1
Genomic Map