Assay Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063947.1 | XP_017919436.1 | 3 - 4 | 695 | 110 | |
| GenBank mRNA | JO334326.2 | - | 1 - 2 | 187 | 110 | |
More Information
Set Membership: |
|

GNA13
Genomic Map