Assay Details
| Gene Symbol: BRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695612.3 | XP_005695669.1 | 2 - 3 | 332 | 73 | |
| GenBank mRNA | JO363185.2 | - | 1 - 2 | 915 | 73 | |
| KA323094.1 | - | 2 - 3 | 278 | 73 | ||
More Information
Set Membership: |
|

BRK1
Genomic Map