Assay Details
| Gene Symbol: ALDH5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696784.3 | XP_005696841.3 | 3 - 4 | 601 | 60 | |
| GenBank mRNA | JO632140.2 | - | 3 - 4 | 482 | 60 | |
| JT675132.1 | - | 10 - 11 | 1519 | 60 | ||
More Information
Set Membership: |
|

ALDH5A1
Genomic Map