Assay Details
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040196.1 | XP_017895685.1 | 4 - 5 | 915 | 64 | |
| XM_018040198.1 | XP_017895687.1 | 4 - 5 | 1013 | 64 | ||
| XM_018040199.1 | XP_017895688.1 | 4 - 5 | 911 | 64 | ||
| XM_018040200.1 | XP_017895689.1 | 4 - 5 | 901 | 64 | ||
More Information
Set Membership: |
|

PTCD1
Genomic Map