Assay Details
| Gene Symbol: PLA2G12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699182.3 | XP_005699239.1 | 3 - 4 | 777 | 61 | |
| XM_018042650.1 | XP_017898139.1 | 2 - 3 | 688 | 61 | ||
| GenBank mRNA | JO413052.2 | - | 2 - 3 | 528 | 61 | |
| JO413053.2 | - | 3 - 4 | 617 | 61 | ||
More Information
Set Membership: |
|

PLA2G12B
Genomic Map