Assay Details
| Gene Symbol: PRKAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962471.2 | XP_013817925.1 | 3 - 4 | 600 | 64 | |
| XM_018045996.1 | XP_017901485.1 | 4 - 5 | 592 | 64 | ||
More Information
Set Membership: |
|

PRKAB2
Genomic Map