Assay Details
| Gene Symbol: LOC102180290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048311.1 | XP_017903800.1 | 9 - 10 | 1215 | 79 | |
| XM_018048312.1 | XP_017903801.1 | 9 - 10 | 1062 | 79 | ||
| XM_018048313.1 | XP_017903802.1 | 9 - 10 | 1002 | 79 | ||
| GenBank mRNA | JO603334.2 | - | 2 - 3 | 552 | 79 | |
More Information
Set Membership: |
|

LOC102180290
Genomic Map