Assay Details
| Gene Symbol: LOC102180290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048311.1 | XP_017903800.1 | 10 - 11 | 1257 | 92 | |
| XM_018048312.1 | XP_017903801.1 | 10 - 11 | 1104 | 92 | ||
| XM_018048313.1 | XP_017903802.1 | 10 - 11 | 1044 | 92 | ||
| GenBank mRNA | JO603334.2 | - | 1 - 2 | 510 | 92 | |
More Information
Set Membership: |
|

LOC102180290
Genomic Map