Assay Details
| Gene Symbol: RHOBTB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050024.1 | XP_017905513.1 | 5 - 6 | 1287 | 66 | |
| GenBank mRNA | KA335569.1 | - | 7 - 8 | 2648 | 66 | |
More Information
Set Membership: |
|

RHOBTB3
Genomic Map