Assay Details
| Gene Symbol: KNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054444.1 | XP_017909933.1 | 13 - 14 | 6066 | 61 | |
| GenBank mRNA | JO593838.2 | - | 13 - 14 | 1550 | 61 | |
More Information
Set Membership: |
|

KNL1
Genomic Map