Assay Details
| Gene Symbol: PHF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056741.1 | XP_017912230.1 | 10 - 11 | 83 | ||
| XM_018056742.1 | XP_017912231.1 | 9 - 10 | 83 | |||
| GenBank mRNA | KA322334.1 | - | 9 - 10 | 814 | 83 | |
More Information
Set Membership: |
|

PHF11
Genomic Map