Assay Details
| Gene Symbol: GALNT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061448.1 | XP_017916937.1 | 7 - 8 | 1736 | 60 | |
| GenBank mRNA | JO644771.2 | - | 4 - 5 | 1318 | 60 | |
More Information
Set Membership: |
|

GALNT9
Genomic Map