Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045026.1 | XP_017900515.1 | 2 - 3 | 255 | 58 | |
| XM_018045027.1 | XP_017900516.1 | 2 - 3 | 257 | 58 | ||
| GenBank mRNA | JO440780.2 | - | 2 - 3 | 820 | 58 | |
More Information
Set Membership: |
|

SNRPD3
Genomic Map