Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056341.1 | XP_017911830.1 | 21 - 22 | 5605 | 68 | |
| GenBank mRNA | KA350996.1 | - | 19 - 20 | 4811 | 68 | |
More Information
Set Membership: |
|

CRYBG2
Genomic Map