Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056341.1 | XP_017911830.1 | 4 - 5 | 3626 | 77 | |
| GenBank mRNA | KA350996.1 | - | 2 - 3 | 2832 | 77 | |
More Information
Set Membership: |
|

CRYBG2
Genomic Map