Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039612.1 | XP_017895101.1 | 2 - 3 | 1982 | 103 | |
XM_018039613.1 | XP_017895102.1 | 2 - 3 | 2154 | 103 | ||
XM_018039614.1 | XP_017895103.1 | 2 - 3 | 1976 | 103 | ||
XM_018039615.1 | XP_017895104.1 | 2 - 3 | 1990 | 103 | ||
XM_018039616.1 | XP_017895105.1 | 2 - 3 | 1982 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
