Assay Details
| Gene Symbol: NUDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040012.1 | XP_017895501.1 | 3 - 4 | 355 | 101 | |
| GenBank mRNA | JO456288.2 | - | 3 - 4 | 337 | 101 | |
More Information
Set Membership: |
|

NUDT1
Genomic Map