Assay Details
| Gene Symbol: LOC102184668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675737.3 | XP_005675794.1 | 5 - 6 | 935 | 91 | |
| XM_018053257.1 | XP_017908746.1 | 5 - 6 | 935 | 91 | ||
More Information
Set Membership: |
|

LOC102184668
Genomic Map