Assay Details
| Gene Symbol: LOC102184668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675737.3 | XP_005675794.1 | 7 - 8 | 1126 | 65 | |
| XM_018053275.1 | XP_017908764.1 | 6 - 7 | 1030 | 65 | ||
More Information
Set Membership: |
|

LOC102184668
Genomic Map