Assay Details
| Gene Symbol: NABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676282.3 | XP_005676339.1 | 5 - 6 | 948 | 102 | |
| XM_005676283.3 | XP_005676340.1 | 5 - 6 | 641 | 102 | ||
More Information
Set Membership: |
|

NABP1
Genomic Map