Assay Details
| Gene Symbol: SMPDL3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056857.1 | XP_017912346.1 | 4 - 5 | 985 | 69 | |
| GenBank mRNA | KA341115.1 | - | 4 - 5 | 1073 | 69 | |
More Information
Set Membership: |
|

SMPDL3B
Genomic Map