Assay Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677142.3 | XP_005677199.3 | 5 - 6 | 955 | 88 | |
| GenBank mRNA | JO503947.2 | - | 1 - 2 | 410 | 88 | |
| JO643382.2 | - | 1 - 2 | 360 | 88 | ||
More Information
Set Membership: |
|

DUSP12
Genomic Map