Assay Details
| Gene Symbol: LOC102186069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677232.3 | XP_005677289.2 | 22 - 23 | 3171 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102186069| Gene Symbol: LOC102186069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677232.3 | XP_005677289.2 | 22 - 23 | 3171 | 118 | |
Set Membership: |
|