Assay Details
| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677657.3 | XP_005677714.2 | 7 - 8 | 1027 | 74 | |
| XM_013962433.2 | XP_013817887.2 | 8 - 9 | 1010 | 74 | ||
| GenBank mRNA | JO580604.1 | - | 1 - 2 | 859 | 74 | |
More Information
Set Membership: |
|

CTSS
Genomic Map