Assay Details
| Gene Symbol: LOC102188370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677751.2 | XP_005677808.1 | 11 - 12 | 1159 | 88 | |
| XM_018046005.1 | XP_017901494.1 | 9 - 10 | 860 | 88 | ||
| GenBank mRNA | JO594747.2 | - | 11 - 12 | 1118 | 88 | |
More Information
Set Membership: |
|

LOC102188370
Genomic Map