Assay Details
| Gene Symbol: LOC102188370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677751.2 | XP_005677808.1 | 12 - 13 | 1255 | 102 | |
| XM_005677753.3 | XP_005677810.1 | 11 - 12 | 1160 | 102 | ||
| XM_018046005.1 | XP_017901494.1 | 10 - 11 | 956 | 102 | ||
| GenBank mRNA | JO594747.2 | - | 12 - 13 | 1214 | 102 | |
More Information
Set Membership: |
|

LOC102188370
Genomic Map