Assay Details
| Gene Symbol: LOC102188370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677751.2 | XP_005677808.1 | 2 - 3 | 258 | 91 | |
| XM_005677753.3 | XP_005677810.1 | 2 - 3 | 259 | 91 | ||
| GenBank mRNA | JO594747.2 | - | 2 - 3 | 217 | 91 | |
| JO646858.2 | - | 8 - 9 | 749 | 91 | ||
More Information
Set Membership: |
|

LOC102188370
Genomic Map