Assay Details
| Gene Symbol: LOC102188370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677751.2 | XP_005677808.1 | 7 - 8 | 776 | 87 | |
| XM_005677753.3 | XP_005677810.1 | 7 - 8 | 777 | 87 | ||
| XM_018046005.1 | XP_017901494.1 | 5 - 6 | 477 | 87 | ||
| GenBank mRNA | JO594747.2 | - | 7 - 8 | 735 | 87 | |
| JO646858.2 | - | 3 - 4 | 231 | 87 | ||
More Information
Set Membership: |
|

LOC102188370
Genomic Map